Learn More
Fine Science Tools Agricola Retractor, Teeth W 5.5 mm, Teeth Depth 4.5 mm, 4 cm L, each
Supplier: Fine Science Tools 1700504
Product: Agricola Retractor - 3x3 Sharp/4cm/3.5cm Maximum Spread/ Teeth: 3x3 Sharp/ Teeth Length: 4.5mm/ Teeth Width: 5.5mm/ Maximum Spread: 3.5cm/ Alloy: Stainless Steel/ Length: 4cm/ Unit: Each