Learn More
Foxx Life Sciences Borosil® Retort Base SS Steel Rod with SS Grade 201, 12 X 500, 2/CS
SDP
Supplier:  Foxx Life Sciences LARS8888500
Encompass_Preferred
Borosil® Retort Base SS Steel Rod is made with SS Grade 201 material. Retort Base Rod can be used with Borosil Heavy Duty Retort base for laboratory application. Availbale in the sizes of 500mm, 600mm and 750mm sizes. In collaboration with Borosil Glass Works, Foxx Life Sciences is proud to bring the outstanding quality of Borosil® Type 3.3 Borosilicate glassware to North America for the first time. Manufactured from the same USP Type 1 borosilicate glass that North American scientists have come to trust, Borosil® Laboratory Glassware offers unparalleled mechanical strength, clarity, and reliability at competitive prices. And thanks to a unique annealing process, Borosil® Glassware is stress-free and incredibly resistant to thermal shock.
Catalog No. 50-172-0523
Explore available promotions