Learn More
Ambu, Inc. Ambu SPUR II Adult Reuscitator | 6/CS
Supplier:  Ambu, Inc. 520611000
Encompass_Preferred
The SPUR II from Ambu provides users with exceptional tactile and visual feedback during resuscitation. The bag walls are thin and responsive, enabling the user to better gauge patient lung compliance during ventilation. Its SafeGrip surface, integrated handle, and lightweight design help to reduce user fatigue. It is made from SEBS material instead of PVC and the bag body’s streamlined design provides optimal stroke volume and superior recoil time compared to other disposable units.
Catalog No. NC1615080
Due to product restrictions, please to purchase or view availability for this product.