Learn More
Restek™ UltraShield™ UHPLC PreColumn Filter
Cost-effective protection for UHPLC systems
Supplier: Restek™ 24997
Description
- In-line design installs easily with standard fittings
- Reliable way to extend column lifetime
- Leak-tight to 15,000 psi (1034 bar)
- 0.5μm titaniun filter in stainless steel body withh PEEK ferrule
Specifications
For Protection From Harmful Effects of Sample and Mobile Phase Particulates on UHPLC Columns | |
Stainless Steel, PEEK (Ferrule) | |
10/Pk. | |
UHPLC PreColumn Filter |