Learn More
Restek™ UltraShield™ UHPLC PreColumn Filter
SDP
Cost-effective protection for UHPLC systems
Supplier:  Restek™ 24996
 View more versions of this product
Catalog No. 06-715-982
Add to Cart
Description
Description
  • In-line design installs easily with standard fittings
  • Reliable way to extend column lifetime
  • Leak-tight to 15,000 psi (1034 bar)
  • 0.5μm titaniun filter in stainless steel body withh PEEK ferrule
Specifications
Specifications
For Protection From Harmful Effects of Sample and Mobile Phase Particulates on UHPLC Columns
Stainless Steel, PEEK (Ferrule)
5/Pk.
UHPLC PreColumn Filter
Product Suggestions
Product Suggestions