Learn More
Thermolite™ Septa from Restek™
SDP
Precision molding ensures consistent, accurate fit
Supplier:  Restek Corporation 27142/DEL
 View more versions of this product
Catalog No. 06-802-722
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
Description
Description
Precision molding ensures consistent, accurate fit
  • Usable to 340°C (644°F) inlet temperature
  • Excellent puncturability
  • Preconditioned and ready to use
  • Packaged in ultra-clean blister packs
Specifications
Specifications
0.43 in.
11 mm
Thermolite
50/Pk.
Product Suggestions
Product Suggestions