Learn More
Restek™ Rxi™5Sil MS Capillary Columns
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Engineered to be a low-bleed GC-MS column
$615.00 - $4430.00
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Diameter (Metric) Inner | Film Thickness | Length (Metric) | Price | Quantity & Availability | |||
06-802-962
![]() |
Restek™
43602 |
0.18 mm | 0.18 μm | 20 m |
Each for $782.00
|
|
|||
06-802-963
![]() |
Restek™
43604 |
0.18 mm | 0.36 μm | 20 m |
Each for $808.00
|
|
|||
06-611-027
![]() |
Restek™
43607 |
0.18 mm | 0.1 μm | 60 m |
Each for $1,475.00
|
|
|||
06-611-026
![]() |
Restek™
43605 |
0.18 mm | 0.18 μm | 40 m |
Each for $1,245.00
|
|
|||
06-611-017
![]() |
Restek™
13608 |
0.25 mm | 0.1 μm | 30 m |
Each for $990.50
|
|
|||
06-611-018
![]() |
Restek™
13623600 |
0.25 mm | 0.25 μm | 30 m |
Pack of 6 for $4,430.00
|
|
|||
06-611-020
![]() |
Restek™
13626 |
0.25 mm | 0.25 μm | 60 m |
Each for $1,391.00
|
|
|||
06-611-021
![]() |
Restek™
13635124 |
0.25 mm | 0.5 μm | 15 m |
Each for $615.00
|
|
|||
06-611-022
![]() |
Restek™
13638124 |
0.25 mm | 0.5 μm | 30 m |
Each for $978.00
|
|
|||
Description
- General-purpose columns
- Excellent inertness for active compounds
- Ideal for GC-MS analysis of semivolatiles, polycyclic aromatic compounds, chlorinated hydrocarbons, phthalates, phenols, amines, organochlorine pesticides, organophosphorus pesticides, drugs, solvent impurities, and hydrocarbons
- Temperature range: -60°C to 350°C
Specifications
Fused Silica |