Learn More
Restek™ RTX™-225 Capillary Columns
Similar to DB-225 and HP-225 columns
Supplier: Restek™ 140206850
Description
- Crossbond™ 50% cyanopropylmethyl/50% phenylmethyl polysiloxane provides low bleed, increased column life, and solvent rinsability
- Ideal confirmation column
- High thermal stability and inertness
- Ideal for the analysis of fatty acid methyl esters (FAMEs), sugar derivatives, and food and flavor analysis
- Include individual test chromatogram
Specifications
Rtx™-225 Column | |
Rtx™-225 | |
40°C to 220/240°C | |
15 m |
0.25 mm | |
Crossbond™ | |
0.25 μm | |
Polar |