Learn More
Restek™ Fittings, Parker (Treated): Tees
![SDP](/content/dam/fishersci/glyphs/sdp.png)
Ideal for installing new equipment, modifying existing instrumentation, or replacing worn connections.
$139.00 - $187.70
Catalog Number | Mfr. No. | Connections | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Connections | Material | Price | Quantity & Availability | ||||
06-711-999
![]() |
Restek™
22528 |
1/4 in. | Sulfinert-coated |
Each for $139.00
|
|
||||
06-711-998
![]() |
Restek™
22527 |
1/8 in. | Sulfinert-coated |
Each for $172.20
|
|
||||
06-711-997
![]() |
Restek™
22526 |
1/16 in. | Sulfinert-coated |
Each for $187.70
|
|
||||
Description
- Available in Siltek/Sulfinert treated or Silcosteel-CR treated fittings
Specifications
Tee |