Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Restek™ Bonded Packed Column Stationary Phases: 20% Rtx-Stabilwax
Bonded phases in capillary columns provide a dramatic increase in column quality. Restek Bonded Packed Column Stationary Phases: 20% Rtx-Stabilwax are for on-column injections.
$237.00
Specifications
Max. Temperature | 240°C |
---|---|
Mesh Size | 100/120 |
Diameter (Metric) Inner | 2 mm |
Diameter (Metric) Outer | 3.17 mm |
Catalog Number | Mfr. No. | Type | Packing Material | Material | Stationary Phase | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Type | Packing Material | Material | Stationary Phase | Price | Quantity & Availability | ||
06-714-604
|
Restek™
80449810 |
Agilent Configuration | Silcoport™ W | Stainless Steel | 20% Rtx™-Stabilwax |
Each for $237.00
|
|
||
06-714-605
|
Restek™
80449851 |
Shimadzu 8A | 20% Rtx™-Stabilwax | Stainless Steel | Bonded Phase 20% Rtx™-Stabilwax (Carbowax™ Phase) |
Each for $237.00
|
|
||
06-714-500
|
Restek™
80417810 |
Agilent Configuration | Silcoport™ W | SilcoSmooth™ | 20% Rtx™-Stabilwax | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
06-714-501
|
Restek™
80417820 |
Varian Configuration | Silcoport™ W | SilcoSmooth™ | 20% Rtx™-Stabilwax | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
06-714-499
|
Restek™
80417800 |
General Configuration | Silcoport™ W | SilcoSmooth™ | 20% Rtx™-Stabilwax | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
06-714-503
|
Restek™
80417840 |
PE Auto Sys Configuration | Silcoport™ W | SilcoSmooth™ | 20% Rtx™-Stabilwax | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
06-714-502
|
Restek™
80417830 |
PE/Sigma Configuration | Silcoport™ W | SilcoSmooth™ | 20% Rtx™-Stabilwax | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
06-714-603
|
Restek™
80449800 |
General Configuration | Silcoport™ W | Stainless Steel | 20% Rtx™-Stabilwax | Item Discontinued This item has been discontinued by the supplier. Please Sign In to view product availability in your area. | N/A | ||
Description
- Low bleed levels
- Longer column lifetimes
- Short conditioning times due to solvent rinsing during manufacturing process which removes low molecular weight polymer fragments
- Bonded packings usually stabilize in 30 minutes or less
- Detectors can be operated at higher sensitivities due to low-bleed
- Reduces need for data system bleed compensation
- More resistant to oxidation than nonbonded phases because of the stronger intermolecular forces produced by cross-linking
- No sagging due to thorough cross-linking of material
- Mesh: 100/120 Silcoport W
Order Info
Initial 2 in. of column will be empty, to accommodate a needle. For a completely filled column add suffix -901. Agilent configuration also includes 1.5 in. void on detector side.
Specifications
240°C | |
100/120 | |
2 mm | |
3.17 mm |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title