Learn More
Hach Company Respirometric BOD Nutrient Buffer Solution Pillows, pk/50
Supplier: Hach Company 2962266
For use with BODTrak II Apparatus. Meets Standard Methods 5210 D requirements. Use one pillow per sample bottle. Included with BODTrak II Apparatus (2952400) and BODTrak II Reagent Set with Bottles (2952440).