Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisher Science Education™ Resorcinol Crystal LG
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
Click to view available options
Quantity:
25 g
Packaging:
Plastic (Code: PGIII)
Specifications
Color | Pink to White |
Assay Percent Range | 1 |
Packaging | Plastic (Code: PGIII) |
Quantity | 25 g |
Formula Weight | 110.112 |
Grade | Laboratory |
Physical Form | Crystals or Flakes |
Provide Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction