Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Eisco™ Resonance Tube Apparatus
SCI_ED

This Resonance Tube Apparatus allows students to determine the speed of sound

Supplier:  Eisco™ PH0720L

Catalog No. S27725


Add to Cart

Description

Description

  • Includes 2 ring clamps
  • Acrylic Tube Dimensions: 47 in. tall, 0.5 in. diameter
  • Metal Base Dimensions: 11 x 10 in., clampable edges
  • Threaded Rod Dimensions: 49 in. long, 0.5 in. diameter
  • Overall Assembled Height: 50 in.
Specifications

Specifications

Resonance Tube Apparatus
2 Ring Clamps
50 in.
Determining The Speed of Sound
1 Ea.
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit