Learn More
Cole-Parmer Myron L CS10 Resistivity Cell, 316SS, K = 0.05, 3/4" NPT(M) thread, thermistor ATC, 10-ft cable
Supplier: Cole-Parmer 0597070
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
Keep your Myron L meter and system running smoothly and accurately with these parts and accessories.
Safety and Handling
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov