Learn More
Electron Microscopy Sciences Light Yellow Melamine Thermosetting Hot Mount Resin 1 KG
Supplier: Electron Microscopy Sciences 1215
Light yellow melamine with mineral and glass filler, thermosetting. For soft to medium hard materials. Excellent edge retention.