Learn More
Serim Research Corp Serim Residual Formaldehyde Test Strips
SDP
Supplier:  Serim Research Corp 5112
Encompass_Preferred
Test measures residual formaldehyde concentrations remaining after rinsing dialyzers or dialysate lines. Sold as one bottle of 100 tests
Catalog No. NC1209952