Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Reserpine, USP, 97-101%, Spectrum™ Chemical
SDP

Catalog Number 18606064
Click to view available options
Quantity:
1 g
5 g
10 g
25 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
50-55-5
C33H40N2O9
608.69
QEVHRUUCFGRFIF-MDEJGZGSSA-N
methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-(3,4,5-trimethoxybenzoyloxy)-3,13-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8-tetraene-19-carboxylate
CO[C@H]1[C@@H](C[C@@H]2CN3CCC4=C(NC5=CC(OC)=CC=C45)[C@H]3C[C@@H]2[C@@H]1C(=O)OC)OC(=O)C1=CC(OC)=C(OC)C(OC)=C1
This item is not returnable. View return policy
50-55-5
C33H40N2O9
608.69
QEVHRUUCFGRFIF-MDEJGZGSSA-N
methyl (1R,15S,17R,18R,19S,20S)-6,18-dimethoxy-17-(3,4,5-trimethoxybenzoyloxy)-3,13-diazapentacyclo[11.8.0.0²,¹⁰.0⁴,⁹.0¹⁵,²⁰]henicosa-2(10),4,6,8-tetraene-19-carboxylate
CO[C@H]1[C@@H](C[C@@H]2CN3CCC4=C(NC5=CC(OC)=CC=C45)[C@H]3C[C@@H]2[C@@H]1C(=O)OC)OC(=O)C1=CC(OC)=C(OC)C(OC)=C1

R3515, 50-55-5, C33H40N2O9

Spectrum™ Chemical Reserpine, USP
Packaging Amber Glass Bottle
Quantity 10 g
CAS Max % 100%
Percent Purity 97 to 101%
Grade USP
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit