Learn More
Eppendorf™ Research™ plus, Mechanical Single Channel Pipettes Multi-Packs PROMO
Eppendorf™ Research™ plus Pipettes meet the highest needs in precision, accuracy, and ergonomics, making them ideal for stress-free work procedures and intuitive operation.
Supplier: Eppendorf™ 13684251S24
Description
- Durable and resistant to chemical, thermal and mechanical stress
- Only a few turns to reach desired volume
- Very low operating force of control button; color indicates pipet volume
- Button is positioned for ideal ergonomics
- Very low operating force of ejector, which is positioned for ideal ergonomics
- Four-digit volume display with magnifying shape
- Secondary adjustment display and opening adjusts pipet to a liquid very different than water
- Eppendorf PerfectPiston™ ultra light piston is made of Fortron™
- Quick connection clip removes lower part easily (not in 10 μl pipettes)
- Spring-loaded tip cone offers improved ergonomics and tight fit to the tip
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Autoclavable | |
Pipette Multipacks | |
Ergonomic |
Research Plus | |
2 to 20, 20 to 200, 100 to 1000 μL | |
Research plus 1-Channel Variable Pipette, 3 epT.I.P.S. Box 2.0, 96 Tips, Ballpoint Pen |