Learn More
Electron Microscopy Sciences Replacement Tip, Straight
Supplier: Electron Microscopy Sciences 7297612
The cautery comes with fully changeable tungsten carbide tips and batteries. Dual power: 1 AA battery for lower power and 2 AA batteries for higher power. Fully autoclavable. Stainless steel construction. Tungsten carbide replacement tip for Low Temperature Cauteryset.