Learn More
Agilent Technologies Replacement onboard Argon bottle. 60 L of welding grade (99.0 %) argon used for plasma ignition with Agilent 4100/4200 MP-AES. Requires – but does NOT include regulator and connecting hose (adaptor kit).
Supplier: Agilent Technologies G800070001
Replacement onboard Argon bottle. A disposable bottle containing 60 L of welding grade (99.0 %) argon used for plasma ignition with Agilent 4100/4200 MP-AES. Requires – but does NOT include regulator and connecting hose (adaptor kit).