Learn More
Hach Company Cl17 Chlorine Analyzer Maintenance Kit (Pre-Assembled Tubing)
Supplier: Hach Company 5444301
Maintenance Kit (for instruments sold after March 2000). Contains pre-assembled reagent tubing, reagent caps and fittings to be replaced annually. Pump module tubing to be replaced at three to six-month intervals.