Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synthware™ Removable Hose Connection Caps
Caps for use with removable hose connectors.
$2.07 - $78.90
Specifications
Color | Red |
---|---|
For Use With (Application) | Organic Synthesis |
Material | Nylon |
Quantity | 1 Ea. |
Catalog Number | Mfr. No. | Thread Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Thread Size | Price | Quantity & Availability | |||||
31-502-299
|
Synthware™
RC50 |
#50 |
Each for $78.90
|
|
|||||
31-501-981
|
Synthware™
RC14 |
#14 |
Each for $2.07
|
|
|||||
31-501-982
|
Synthware™
RC18 |
#18 |
Each for $4.36
|
|
|||||
31-502-300
|
Synthware™
RC7 |
#7 |
Each for $2.80
|
|
|||||
Description
- For use with removable hose connections
- Available in a wide variety of sizes
Specifications
Red | |
Organic Synthesis | |
Nylon | |
1 Ea. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title