Learn More
BrandTech™ Remote Dispensing System and Accessories for BRAND Dispensette Bottletop Dispensers
Accessories to facilitate remote dispensing with a BRAND Dispensette S or Dispensette S Organic bottletop dispenser. Dispenser not included
Supplier: BrandTech™ 704267
Description
- 704261- Remote Dispensing System for Dispensette. Includes wall mount
- 704267 - 10 meters of filling tube
- 704269 - 1 meter filling tube
Specifications
FEP Tubing | |
Translucent | |
Remote dispensing | |
10 m of tubing | |
10 m |
Not Autoclavable | |
FEP | |
Dispensette™ | |
Non-sterile |