Learn More
Thermo Scientific™ Remel™ Xpect™ Giardia Control Kit
Use in qualitative procedures as external control material for the Xpect Giardia or Xpect Giardia/Cryptosporidium test kits.
Supplier: Thermo Scientific™ R2450030
Description
Remel Xpect Giardia Control Kit is used in qualitative procedures as external control material for the Xpect Giardia or Xpect Giardia/Cryptosporidium test kits.
Includes:
1 vial positive control and 1 vial negative control
Specifications
Xpect Flu A and B Sample Panel | |
Giardia Control Kit | |
18 Vials/Pk. |
Giardia | |
12 positive vials and 6 negative vials |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.