Learn More
Thermo Scientific™ Remel™ XLD Agar (Xylose, Lysine, Desoxycholate)
Thermo Scientific™ Remel XLD Agar (Xylose, Lysine, Desoxycholate) is for the selective and differential isolation of gram-negative enteric pathogens.
Supplier: Thermo Scientific™ R01981
This item is not returnable.
View return policy
Description
Thermo Scientific™ Remel XLD Agar (Xylose, Lysine, Desoxycholate) is for the selective and differential isolation of gram-negative enteric pathogens.
Specifications
Industrial Reference: AOAC, BAM, COMPF, USP. | |
10mm x 100mm Spacesaver Plate | |
15/Pk. |
XLD Agar | |
Agar |