Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Thermo Scientific™ Remel ProSpecT™ Cryptosporidium Rapid Assay Sample Panel
Supplier: Thermo Scientific™ R2448035
This item is not returnable.
View return policy
Description
- For use with ProSpect Cryptosporidium Rapid Assay
- Contains 2 × 0.75mL positive vials and 2 × 0.75mL negative vials
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction