Learn More
Thermo Scientific™ Phenylethyl Alcohol Agar with 5% Sheep Blood
Isolate and cultivate Gram-positive cocci, such as staphylococci and streptococci, from specimens containing mixed flora with this selective medium
Supplier: Thermo Scientific™ R01662
Description
Isolate and cultivate Gram-positive cocci, such as staphylococci and streptococci, from specimens containing mixed flora1 with Thermo Scientific™ Remel™ Phenylethyl Alcohol Agar with 5% Sheep Blood. This medium permits the growth of Gram-positive organisms while inhibiting most Gram-negative organisms, especially the swarming of Proteus species.
Phenylethyl Alcohol Agar with 5% Sheep Blood is formulated as per Lilley and Brewer1 for selective isolation of Gram-positive bacteria. This medium is especially useful when specimens are contaminated with swarming Proteus species.
- Selective: phenylethyl alcohol in the medium inhibits Gram-negative organisms by inhibiting their DNA synthesis.
- Ready-to-use: convenience of prepared media.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
Phenylethyl Alcohol Agar w/5% Sheep Blood | |
12mm x 85mm Monoplate | |
Agar | |
100/Pk. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.