Learn More
Thermo Scientific™ PathoDx™ Herpes Typing Control Slide
PathoDx™ and Herpes Typing Control Slide is used with Herpes immunofluorescent assays.
Supplier: Thermo Scientific™ R62251
Description
Thermo Scientific™ Remel™ PathoDx™ Herpes Typing Control Slide is used with Herpes immunofluorescent assays. Herpes Simplex Virus (HSV) is an ancient and ubiquitous virus, known to cause acute and recurrent infections in humans. In the 1960s, it was recognized that HSV consisted of two distinct types, HSV-I and HSV-II1. HSV-I was considered to be primarily associated with ocular and oral infection (above the waist) while HSV-II was considered to be a genital infection (below the waist).
PathoDx Herpes Typing Kit uses four fluorescein-labeled monoclonal antibodies specific for HSV-I and HSV-II (two for HSV-I; two for HSV-II) to detect and identify Herpes Simplex Virus. The standard method for diagnosis of HSV has been growth in tissue culture cells2,3. The reagents are specific for each HSV type and will not show any cross-reactive staining.
- Ready-to-use – In vitro diagnostic medical device
- Easy-to-store – Store at 2-8°C
Herpes Typing Control Slides are five individually foil-sealed slides packaged in a zip lock bag with desiccant. Each control slide has four wells. The two upper wells contain RK-13 cells infected with HSV-I (strain MacIntyre), and the two lower wells contain RK-13 cells infected with HSV-II (strain MS).
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
Specifications
PathoDX™ Herpes Typing Control Slide | |
5 Slides, Each slide has 4 wells; 2 containing HSV-I infected cells and 2 containing HSV-II infected cells | |
5 Slides/Pk. |
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.