Learn More
Thermo Scientific™ PathoDx Chlamydia Control Slide
Use Chlamydia Control Slides with Chlamydia Direct Specimen Kit for immunofluorescence assays.
Supplier:  Thermo Scientific™ R62205
Catalog No. R62205
Add to Cart
This item is not returnable. View return policy
Description
Description
Use Thermo Scientific™ Remel™ PathoDx™ Chlamydia Control Slides for direct immunofluorescence assays of Chlamydia. PathoDx™ Chlamydia Control Slides are intended for use with the Chlamydia Direct Specimen kit (R62220), which contains all components necessary for the test system. These slides are provided as replacement items.
Chlamydia Control Slides contain five slides packaged individually in sealed foil with desiccant. Each control slide has two wells - one positive and one negative.
The positive well of the control slide contains uninfected fixed McCoy cells and inactivated C. trachomatis elementary bodies. The negative well contains only uninfected McCoy cells.
The slide should be warmed to room temperature (15 to 28°C) before use while still in its foil package.
Not all products are available for sale in all territories. Please inquire.
Remel™ and Oxoid™ products are now part of the Thermo Scientific brand.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
PathoDX™ Chlamydia Control Slide
Chlamydia
Chlamydia Control Slide
5 Slides/Pk.