Learn More
Thermo Scientific™ Remel™ Fluid A
Thermo Scientific™ Remel Fluid A with 0.1% peptone (Peptone Fluid) is used as diluting or rinsing fluid for sterility testing.
Supplier: Thermo Scientific™ R112302
This item is not returnable.
View return policy
Description
Thermo Scientific™ Remel Fluid A with 0.1% peptone (Peptone Fluid) is used as diluting or rinsing fluid for sterility testing.
Specifications
Fluid A | |
Septum - 500mL | |
Buffer Solution | |
10 x 500 mL |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Not intended for IVD use.