Learn More
Thermo Scientific™ Remel™ BactiSwab™ Gel Collection and Transport Swabs: Single Swab Amies Charcoal
Thermo Scientific™ Remel BactiSwab Gel Collection and Transport Swabs provide excellent recovery for a wide range of bacteria, including anaerobes.
$120.00 - $949.00
Specifications
Format | Tube |
---|---|
Media Type | Amies Charcoal |
Closure Color | Black |
Description | Amies Charcoal |
Quantity | 1 Tube/Envelope |
Description
Thermo Scientific™ Remel BactiSwab Gel Collection and Transport Swabs provide excellent recovery for a wide range of bacteria, including anaerobes.
Rayon-tipped, plastic-shaft swab(s) are mounted in the cap of a translucent plastic tube with spiral chamber containing gel transport medium with or without charcoal.- Recommended for recovery of Neisseria gonorrhoeae
- Single plastic shaft
- Black cap
- Round-bottom tube for easy racking and seamless molding to ensure strength and integrity
- Exclusive zip-seal packaging to promote optimum media performance
- Color-coded cap with protective overskirt
- Inert, non-toxic rayon swab tip for good sample retrieval and absorption
Specifications
Tube | |
Black | |
1 Tube/Envelope |
Amies Charcoal | |
Amies Charcoal | |
Plastic |