Learn More
Thermo Scientific™ Remel™ Agglutinating Sera, Haemophilus influenzae
Used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
Supplier:  Thermo Scientific™ R30166201
 View more versions of this product
Catalog No. R30166201
Add to Cart
This item is not returnable. View return policy
Description
Description
Remel Agglutinating Sera are used for serological identification of H. influenzae by rapid slide agglutination test and CIE procedures.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Agglutinating Sera, Haemophilus influenzae Type C
H. influenzae
Agglutinating Sera