Learn More
Medchemexpress LLC HY-104077 10mg Medchemexpress, Remdesivir CAS:1809249-37-3 Purity:>98%
Supplier: Medchemexpress LLC HY10407710MG
Medchemexpress, HY-104077 10mg Remdesivir CAS:1809249-37-3 Remdesivir (GS-5734), a nucleoside analogue with effective antiviral activity, has EC50s of 74 nM for SARS-CoV and MERS-CoV in HAE cells, and 30 nM for murine hepatitis virus in delayed brain tumor cells. Remdesivir is highly effective in the control of SARS-CoV-2 (COVID-19) infection in vitro[2]. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.