Learn More
All Safe Industries REGULATOR, PRESET FLOW TO 1.0
SDP
Supplier:  All Safe Industries AS17151.0
Encompass_Preferred
Regulator, Preset Flow to 1.0 LPM. Fits 8AL, 6D and 2AL Cylinders
Catalog No. NC2041553