Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Refrigerator/Freezer Storage Racks
![SCI_ED](/content/dam/fishersci/glyphs/scied.png)
For sample storage in refrigerator or freezer.
Description
- Clear acrylic racks store 16 mm O.D. specimen tubes in refrigerator or freezer
- Allow sufficient space between tubes to let you read individual tube labels
- Make efficient use of shelf space
- Handgrips for easy carrying
Order Info
Additional rack sizes available. To order, contact your Fisher Scientific Customer Service Representative.
Specifications
Refrigerator/Freezer Rack | |
Transparent | |
Acrylic | |
100 | |
24.1 cm | |
24.1 cm | |
6.3 cm | |
16 mm |
Not Autoclavable | |
Tubes (16 mm) | |
100 | |
9.5 x 9.5 x 2.5 in. (24.1 x 24.1 x 6.3 cm) | |
9.5 in. | |
9.5 in. | |
2.5 in. |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction