Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Reformulated Organohalide Pesticides Mix, SPEX CertiPrep™
ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material
Supplier: SPEX CertiPrep 505A2
Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Description
Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation
Analyte | CAS No. | Concentration |
---|---|---|
Atrazine | 1912-24-9 | 1250μg/mL |
Simazine | 122-34-9 | 1250μg/mL |
Alachlor | 15972-60-8 | 50μg/mL |
Methoxychlor | 72-43-5 | 25μg/mL |
Aldrin | 309-00-2 | 5μg/mL |
alpha-Chlordane | 5103-71-9 | 5μg/mL |
cis-Nonachlor | 5103-73-1 | 5μg/mL |
Dieldrin | 60-57-1 | 5μg/mL |
Endrin | 72-20-8 | 5μg/mL |
gamma-BHC | 58-89-9 | 5μg/mL |
gamma-Chlordane | 5103-74-2 | 5μg/mL |
Heptachlor | 76-44-8 | 5μg/mL |
Heptachlor epoxide (Isomer B) | 1024-57-3 | 5μg/mL |
Hexachlorobenzene | 118-74-1 | 5μg/mL |
Hexachlorocyclopentadiene | 77-47-4 | 5μg/mL |
trans-Nonachlor | 39765-80-5 | 5μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Reformulated Organohalide Pesticides Mix | |
2mL amber prescored glass ampule | |
Acetone | |
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation |
USEPA Method 505 | |
For US EPA Method 505 | |
1 mL | |
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity. |
Safety and Handling
DOTInformation : Nonhazardous
Recommended Storage : Freezer
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Product intended for laboratory use only
Spot an opportunity for improvement?
Provide Content Correction