Learn More
Cole-Parmer Monarch MI153209 Roll of Reflective Tape for use with Optical Tachometers, 5 ft long and 1/2" wide
Supplier: Cole-Parmer 0821062
Cut and adhere this reflective tape to the surface being monitored.