Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Zoll Medical Refill Nitrile Gloves for Mobilize Trauma Kits
Description
Note: Serial number of the trauma kit this is being ordered for is required upon order.
Specifications
Color | Blue |
Disposable | Single-use |
Material | Nitrile |
Product Type | Gloves |
Size | Large |
For Use With (Equipment) | Mobilize Trauma Kits |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction