Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Fisherbrand™ Redi-Tip™ General Purpose Pipet Tips

High-quality tips made of premium virgin polypropylene resin using advanced molding methods
Description
- Certified by independent laboratory
- Nonsterile: RNase, DNase, DNA, and pyrogen free
- Sterile: RNase, DNase, DNA, Bioburden, PCR inhibitor and pyrogen free
- For use with most 200 and 1000 μL single and multichannel pipettors
- Hinged racks can be stacked rack-on-rack
- Hinged closure on racks allows top lid to be partially open for steam penetration during autoclaving
Specifications
100 to 1000 μL | |
Blue | |
2.75 in. | |
DNase, RNase, DNA and pyrogen free | |
Universal | |
Polypropylene |
Non-sterile | |
Autoclavable | |
6.9 cm | |
Hinged Rack | |
Universal fit pipettor |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title