Learn More
Electron Microscopy Sciences Brush, Red Sable, Size #5/0, 1 ea
Supplier: Electron Microscopy Sciences 6610050
Extremely soft and pliable. Perfect for the manipulation of previously fixed, dehydrated specimens into embedding capsules or molds.