Learn More
StatLab Red Line Plus Adhesion Control Slide
![SDP](/content/dam/fishersci/glyphs/sdp.png)
A high-quality alternative to expensive brand name adhesion control slides.
Supplier: StatLab ES355PLUS
Description
- Features a red line printed on the bottom of each slide to designate areas for patient tissue and positive controls
- A high quality alternative to expensive brand name control slides
- These adhesion slides are provided without control tissue
- 144 slides per gross
Specifications
Control Slide |