Learn More
3M™ Red Buffer Pad
3M™ Red Buffer Pads are especially designed for spray buffing. Cleans when damp; buffs when dry. Quickly cleans and removes scuff marks and enhances floor appearance. Works great on automatic floor scrubbers for light duty cleaning.
Supplier: 3M™ 7000000680
This item is not returnable.
View return policy
Description
- Use to light scrub without dulling the finish
- Can be used for everyday cleaning under and automatic scrubber
- Also designed for spray buffing
- Cleans when damp; buffs when dry
- Use on rotary or automatic floor machines
- *Certified by Green Seal™ for Environmental Innovation based on a design for significantly improved initial performance compared to competitive products, extending the useful life of the floor pad and reducing solid waste generation
- *Certified by Green Seal™ for Environmental Innovation based on a design for significantly improved initial and mid-life performance compared to competitive products, extending the useful life of the floor pad and reducing solid waste generation
Specifications
Cleaning Pad | |
406 mm | |
Rotary or Automatic Floor Machines | |
5100 |
Red | |
Buffing, Cleaning | |
Non-Woven Polyester Fiber | |
5/Cs. |