Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
United Scientific Red Banana/Alligator Cord
Banana/Alligator Cord for use with multiple electricity demonstrations.
$2.70 - $3.70
Specifications
For Use With (Application) | Electricity Demonstration |
---|---|
Product Type | Wires |
Color | Red |
Description
- Flexible, 18 gauge (SWG) stranded wire cord with a stackable banana plug at one end and an alligator clip at the other end
- Alligator clip has a protective vinyl sheath
- 12 in., 24 in., or 36 in. lengths
- Prop 65 Warning Label
Specifications
Electricity Demonstration | |
Wires | |
Red |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title