Learn More
Fisherbrand™ Rectangular HDPE Dipping Baskets
SDPSCI_ED
Convenient rectangular shape easily fits into standard tanks.
 View more versions of this product
Catalog No. S37140
Add to Cart
Description
Description
  • High density polyethylene with 1/8 in. thick walls, 3/16 in. diameter perforations on 5/16 in. centers
  • Excellent for washing, pickling or dipping into corrosive liquids
  • Supplied with perforated bottoms and sides and solid ends
Order Info
Additional dipping basket sizes available. To order, contact your Fisher Scientific Customer Service Representative.
Specifications
Specifications
Not Autoclavable
HDPE
30.48 cm
20.32 cm
15.2 cm
0.45 cm
White
12 x 8 x 6 in. (30.48 x 20.32 x 15.2 cm)
12 in.
8 in.
6 in.
Product Suggestions
Product Suggestions