Learn More
Fisherbrand™ Rectangular, Epoxy-Coated Test Tube Basket
SDP
These Epoxy-Coated Test Tube Baskets are useful in holding laboratory ware for washing, storage, or autoclaving.
 View more versions of this product
Catalog No. 01-189-111
Add to Cart
Description
Description
  • Useful in holding laboratory ware for washing, storage, or autoclaving
  • Epoxy-coated for non-stick applications
  • Flow-through design
  • Autoclavable and durable
  • Withstands temperatures from -220°C to 120°C
Specifications
Specifications
Basket
Autoclavable
Nylon-coated Stainless Steel
Yes
Test tubes and other labwares
ISO, GMP
White
Rectangle
Non-sterile
6 x 4 x 4 in.
Product Suggestions
Product Suggestions