Learn More
Cole-Parmer Cole-Parmer 2 Channel Electronic Paperless Recorder, 100 to 240 VAC
Supplier:  Cole-Parmer 8081600
Encompass_Preferred
The most economical two-channel electronic recorder on the market! View data in dual trend, dual digital, digital inversed, or split screened formats. User-friendly interface makes configuration and setup simple.
Catalog No. 50-631-716