Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
R&D Systems™ Recombinant Mouse Wnt-3a Protein
Extensive quality control produces industry leading bioactivity and lot-to-lot consistency that instills confidence in results and ensures reproducibility. Applications: Bioactivity
$328.28 - $1026.60
Specifications
Gene ID (Entrez) | 22416 |
---|---|
Molecular Weight (g/mol) | 37 kDa |
Name | Wnt-3a |
Source | Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-3a protein Ser19-Lys352 |
Storage Requirements | Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. |
Catalog Number | Mfr. No. | Formulation | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Formulation | Quantity | Price | Quantity & Availability | ||||
1324WN002
|
R&D Systems™
1324WN002 |
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. | 2μg |
Each for $328.28
|
|
||||
1324WN002CF
|
R&D Systems™
1324WN002/CF |
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. | 2μg |
Each for $328.28
|
|
||||
1324WN010
|
R&D Systems™
1324WN010 |
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS with BSA as a carrier protein. | 10μg |
Each for $1,026.60
|
|
||||
1324WN010CF
|
R&D Systems™
1324WN010/CF |
Lyophilized from a 0.2μm filtered solution in PBS, EDTA and CHAPS. | 10μg |
Each for $1,026.60
|
|
||||
Description
The Recombinant Mouse Wnt-3a Protein is derived from CHO. The Recombinant Mouse Wnt-3a Protein has been validated for the following applications: Bioactivity.Specifications
22416 | |
Wnt-3a | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70° C as supplied. 1 month, 2 to 8° C under sterile conditions after reconstitution. 3 months, -20 to -70° C under sterile conditions after reconstitution. | |
Recombinant |
37 kDa | |
Chinese Hamster Ovary cell line,CHO-derived mouse Wnt-3a protein Ser19-Lys352 | |
Unconjugated |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title