Learn More
Electron Microscopy Sciences Light Pipe Scintillator Recoating Surface
Supplier: Electron Microscopy Sciences 82008RS
![Encompass_Preferred](/content/dam/fishersci/glyphs/encompass_preferred.png)
The light pipe is the connection between the scintillator and the phosphomultiplier tube (PMT). It transfers the photons produced by the scintillator for detection by the PMT. We coat the scintillator material directly on the light pipe which improves the signal. Price includes one Re-coating Certificate. Re-coating is typically needed twice per year. Typically needed twice per year. Price includes one recoating certificate
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.