Learn More
Perkin Elmer US LLC Rechargeable Desiccant Kit for Spectrum and Paragon Spectrometers
Supplier:  Perkin Elmer US LLC L1180480
Encompass_Preferred
Rechargeable Desiccant Kit comes with two desiccant cartridges. Rechargeable desiccant cartridges are contained in rigid metal cages which are user-installable. One cartridge is required for the 1000/RX/ BX and Paragon 500/1000/1600 instruments. The second cartridge is a spare. It is recommended to have spare desiccant cartridges available so when a desiccant change is required recharged desiccant can be installed immediately.
Catalog No. 50-210-3696