Learn More
Perkin Elmer US LLC Rechargeable Desiccant Kit for Spectrum 400
Supplier:  Perkin Elmer US LLC L1251840
Encompass_Preferred
Dessicant packs can be reactivated by baking them in an oven at 250°C for 8 hours and then cooling them in a dry atmosphere. For optimal results, re-activate the packets immediately before re-use.
Catalog No. 50-211-3596